Assay Details
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040766.1 | XP_017896255.1 | 11 - 12 | 1077 | 66 | |
| XM_018040767.1 | XP_017896256.1 | 11 - 12 | 1163 | 66 | ||
More Information
Set Membership: |
|

CCZ1
Genomic Map