Assay Details
| Gene Symbol: WNT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038229.1 | XP_017893718.1 | 3 - 4 | 2519 | 78 | |
| XM_018038230.1 | XP_017893719.1 | 4 - 5 | 1786 | 78 | ||
| XM_018038231.1 | XP_017893720.1 | 3 - 4 | 1721 | 78 | ||
| GenBank mRNA | JO403795.2 | - | 1 - 2 | 224 | 78 | |
More Information
Set Membership: |
|

WNT5A
Genomic Map