Assay Details
| Gene Symbol: ST8SIA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974425.2 | XP_013829879.1 | 4 - 5 | 780 | 61 | |
| XM_013974426.2 | XP_013829880.1 | 3 - 4 | 678 | 61 | ||
| GenBank mRNA | JT670316.1 | - | 7 - 8 | 1757 | 61 | |
More Information
Set Membership: |
|

ST8SIA5
Genomic Map