Assay Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047126.1 | XP_017902615.1 | 2 - 3 | 941 | 71 | |
| XM_018047127.1 | XP_017902616.1 | 3 - 4 | 1050 | 71 | ||
| XM_018047128.1 | XP_017902617.1 | 2 - 3 | 586 | 71 | ||
| GenBank mRNA | JO496372.2 | - | 2 - 3 | 818 | 71 | |
More Information
Set Membership: |
|

KBTBD2
Genomic Map