Assay Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047126.1 | XP_017902615.1 | 3 - 4 | 1111 | 101 | |
| XM_018047127.1 | XP_017902616.1 | 4 - 5 | 1220 | 101 | ||
| XM_018047128.1 | XP_017902617.1 | 3 - 4 | 756 | 101 | ||
| GenBank mRNA | JO496372.2 | - | 3 - 4 | 988 | 101 | |
| JT732206.1 | - | 3 - 4 | 753 | 101 | ||
More Information
Set Membership: |
|

KBTBD2
Genomic Map