Assay Details
| Gene Symbol: SGCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683219.3 | XP_005683276.1 | 7 - 8 | 940 | 96 | |
| XM_013965722.2 | XP_013821176.1 | 10 - 11 | 1979 | 96 | ||
| XM_013965725.2 | XP_013821179.1 | 9 - 10 | 1839 | 96 | ||
| XM_018050121.1 | XP_017905610.1 | 10 - 11 | 1924 | 96 | ||
More Information
Set Membership: |
|

SGCD
Genomic Map