Assay Details
| Gene Symbol: EXOSC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696051.3 | XP_005696108.1 | 7 - 8 | 816 | 69 | |
| GenBank mRNA | JO612047.2 | - | 5 - 6 | 616 | 69 | |
| JO612048.2 | - | 7 - 8 | 813 | 69 | ||
More Information
Set Membership: |
|

EXOSC7
Genomic Map