Assay Details
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062782.1 | XP_017918271.1 | 6 - 7 | 997 | 70 | |
XM_018062783.1 | XP_017918272.1 | 7 - 8 | 847 | 70 | ||
XM_018062784.1 | XP_017918273.1 | 7 - 8 | 814 | 70 | ||
XM_018062785.1 | XP_017918274.1 | 7 - 8 | 793 | 70 | ||
XM_018062786.1 | XP_017918275.1 | 7 - 8 | 760 | 70 | ||
GenBank mRNA | JO641907.2 | - | 1 - 2 | 103 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
