Assay Details
| Gene Symbol: ZNF185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044711.1 | XP_017900200.1 | 13 - 14 | 1351 | 59 | |
| XM_018044712.1 | XP_017900201.1 | 13 - 14 | 988 | 59 | ||
| XM_018044713.1 | XP_017900202.1 | 13 - 14 | 1348 | 59 | ||
| XM_018044714.1 | XP_017900203.1 | 13 - 14 | 1348 | 59 | ||
| XM_018044715.1 | XP_017900204.1 | 13 - 14 | 1345 | 59 | ||
| XM_018044716.1 | XP_017900205.1 | 12 - 13 | 1275 | 59 | ||
| XM_018044717.1 | XP_017900206.1 | 12 - 13 | 1272 | 59 | ||
| XM_018044718.1 | XP_017900207.1 | 13 - 14 | 1351 | 59 | ||
| GenBank mRNA | KA311476.1 | - | 12 - 13 | 1222 | 59 | |
More Information
Set Membership: |
|

ZNF185
Genomic Map