Assay Details
| Gene Symbol: GLDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054194.1 | XP_017909683.1 | 8 - 9 | 1040 | 62 | |
| XM_018054196.1 | XP_017909685.1 | 7 - 8 | 1022 | 62 | ||
| GenBank mRNA | JO426669.1 | - | 2 - 3 | 1961 | 62 | |
More Information
Set Membership: |
|

GLDN
Genomic Map