Assay Details
| Gene Symbol: CCDC113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692042.2 | XP_005692099.1 | 5 - 6 | 698 | 67 | |
| XM_005692043.2 | XP_005692100.1 | 5 - 6 | 620 | 67 | ||
| XM_005692044.2 | XP_005692101.1 | 4 - 5 | 458 | 67 | ||
More Information
Set Membership: |
|

CCDC113
Genomic Map