Assay Details
| Gene Symbol: ANKRD54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048740.1 | XP_017904229.1 | 3 - 4 | 603 | 53 | |
| GenBank mRNA | JT644457.1 | - | 6 - 7 | 466 | 53 | |
| JT689793.1 | - | 10 - 11 | 1659 | 53 | ||
More Information
Set Membership: |
|

ANKRD54
Genomic Map