Assay Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691223.3 | XP_005691280.1 | 8 - 9 | 776 | 68 | |
| XM_013970518.2 | XP_013825972.1 | 8 - 9 | 645 | 68 | ||
| XM_018061561.1 | XP_017917050.1 | 8 - 9 | 785 | 68 | ||
| GenBank mRNA | JT669026.1 | - | 2 - 3 | 227 | 68 | |
More Information
Set Membership: |
|

ZNF330
Genomic Map