Assay Details
| Gene Symbol: SLC13A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693253.3 | XP_005693310.3 | 11 - 12 | 1805 | 61 | |
| XM_013971950.2 | XP_013827404.2 | 11 - 12 | 1611 | 61 | ||
More Information
Set Membership: |
|

SLC13A2
Genomic Map