Assay Details
| Gene Symbol: TMEM38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052554.1 | XP_017908043.1 | 4 - 5 | 648 | 65 | |
| GenBank mRNA | JO583845.1 | - | 4 - 5 | 619 | 65 | |
| JT621067.1 | - | 4 - 5 | 565 | 65 | ||
More Information
Set Membership: |
|

TMEM38B
Genomic Map