Assay Details
| Gene Symbol: SPCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041987.1 | XP_017897476.1 | 4 - 5 | 1727 | 105 | |
| GenBank mRNA | JO344421.2 | - | 1 - 2 | 847 | 105 | |
| JP351143.2 | - | 4 - 5 | 1063 | 105 | ||
More Information
Set Membership: |
|

SPCS3
Genomic Map