Assay Details
| Gene Symbol: IL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686667.3 | XP_005686724.2 | 4 - 5 | 406 | 67 | |
| XM_005686668.3 | XP_005686725.2 | 4 - 5 | 406 | 67 | ||
| XM_005686669.2 | XP_005686726.2 | 4 - 5 | 396 | 67 | ||
| XM_013967721.2 | XP_013823175.2 | 4 - 5 | 396 | 67 | ||
| GenBank mRNA | KA329633.1 | - | 2 - 3 | 675 | 67 | |
More Information
Set Membership: |
|

IL37
Genomic Map