Assay Details
Gene Symbol: IL37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686667.3 | XP_005686724.2 | 5 - 6 | 552 | 91 | |
XM_005686669.2 | XP_005686726.2 | 5 - 6 | 542 | 91 | ||
GenBank mRNA | KA329633.1 | - | 1 - 2 | 529 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
