Assay Details
Gene Symbol: FAM155A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687778.3 | XP_005687835.3 | 1 - 2 | 1398 | 66 | |
XM_018056370.1 | XP_017911859.1 | 1 - 2 | 1398 | 66 | ||
GenBank mRNA | JO634628.2 | - | 1 - 2 | 623 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
