Assay Details
| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061879.1 | XP_017917368.1 | 5 - 6 | 901 | 118 | |
| XM_018061880.1 | XP_017917369.1 | 5 - 6 | 901 | 118 | ||
| XM_018061881.1 | XP_017917370.1 | 4 - 5 | 732 | 118 | ||
| GenBank mRNA | KA326469.1 | - | 5 - 6 | 912 | 118 | |
More Information
Set Membership: |
|

SLC22A31
Genomic Map