Assay Details
| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061879.1 | XP_017917368.1 | 6 - 7 | 1009 | 68 | |
| GenBank mRNA | KA326469.1 | - | 6 - 7 | 1020 | 68 | |
More Information
Set Membership: |
|

SLC22A31
Genomic Map