Assay Details
| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061879.1 | XP_017917368.1 | 7 - 8 | 1227 | 59 | |
| XM_018061880.1 | XP_017917369.1 | 7 - 8 | 1137 | 59 | ||
| XM_018061881.1 | XP_017917370.1 | 6 - 7 | 1071 | 59 | ||
| GenBank mRNA | KA326469.1 | - | 7 - 8 | 1238 | 59 | |
More Information
Set Membership: |
|

SLC22A31
Genomic Map