Assay Details
| Gene Symbol: SEC61G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695496.3 | XP_005695553.1 | 2 - 3 | 372 | 101 | |
| GenBank mRNA | JO580273.1 | - | 2 - 3 | 421 | 101 | |
More Information
Set Membership: |
|

SEC61G
Genomic Map