Assay Details
| Gene Symbol: FAM222A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061104.1 | XP_017916593.1 | 1 - 2 | 499 | 96 | |
| GenBank mRNA | JO621180.2 | - | 2 - 3 | 1228 | 96 | |
| JO621181.2 | - | 2 - 3 | 1228 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018061104.1 transcripts.
Set Membership: |
|

FAM222A
Genomic Map