Assay Details
| Gene Symbol: ARRDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056202.1 | XP_017911691.1 | 5 - 6 | 690 | 68 | |
| XM_018056203.1 | XP_017911692.1 | 5 - 6 | 688 | 68 | ||
| XM_018056204.1 | XP_017911693.1 | 4 - 5 | 639 | 68 | ||
More Information
Set Membership: |
|

ARRDC1
Genomic Map