Assay Details
| Gene Symbol: EFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066294.1 | XP_017921783.1 | 12 - 13 | 1412 | 64 | |
| XM_018066295.1 | XP_017921784.1 | 12 - 13 | 1405 | 64 | ||
More Information
Set Membership: |
|

EFL1
Genomic Map