Assay Details
| Gene Symbol: TEX264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695907.3 | XP_005695964.1 | 3 - 4 | 725 | 62 | |
| XM_013973645.2 | XP_013829099.1 | 3 - 4 | 452 | 62 | ||
| XM_018038292.1 | XP_017893781.1 | 3 - 4 | 450 | 62 | ||
| XM_018038293.1 | XP_017893782.1 | 3 - 4 | 717 | 62 | ||
| XM_018038294.1 | XP_017893783.1 | 3 - 4 | 446 | 62 | ||
| XM_018038295.1 | XP_017893784.1 | 1 - 2 | 382 | 62 | ||
| GenBank mRNA | JO611755.2 | - | 3 - 4 | 479 | 62 | |
More Information
Set Membership: |
|

TEX264
Genomic Map