Assay Details
| Gene Symbol: AMDHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680468.3 | XP_005680525.2 | 5 - 6 | 901 | 58 | |
| GenBank mRNA | JO412624.2 | - | 4 - 5 | 869 | 58 | |
More Information
Set Membership: |
|

AMDHD1
Genomic Map