Assay Details
| Gene Symbol: NINJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681001.3 | XP_005681058.2 | 3 - 4 | 773 | 69 | |
| GenBank mRNA | KA315615.1 | - | 3 - 4 | 606 | 69 | |
More Information
Set Membership: |
|

NINJ2
Genomic Map