Assay Details
| Gene Symbol: TGM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688223.3 | XP_005688280.2 | 7 - 8 | 1145 | 60 | |
| XM_005688224.3 | XP_005688281.2 | 6 - 7 | 1016 | 60 | ||
| GenBank mRNA | KA346110.1 | - | 7 - 8 | 1086 | 60 | |
More Information
Set Membership: |
|

TGM6
Genomic Map