Assay Details
| Gene Symbol: TMEM219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697681.3 | XP_005697738.1 | 3 - 4 | 1649 | 61 | |
| XM_005697682.3 | XP_005697739.1 | 4 - 5 | 1104 | 61 | ||
| XM_005697683.3 | XP_005697740.1 | 4 - 5 | 1101 | 61 | ||
| XM_018039949.1 | XP_017895438.1 | 4 - 5 | 752 | 61 | ||
| XM_018039950.1 | XP_017895439.1 | 4 - 5 | 744 | 61 | ||
| XM_018039951.1 | XP_017895440.1 | 5 - 6 | 818 | 61 | ||
| XM_018039952.1 | XP_017895441.1 | 5 - 6 | 814 | 61 | ||
| GenBank mRNA | JO329880.2 | - | 3 - 4 | 1096 | 61 | |
| KA348803.1 | - | 2 - 3 | 290 | 61 | ||
More Information
Set Membership: |
|

TMEM219
Genomic Map