Assay Details
| Gene Symbol: RAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040404.1 | XP_017895893.1 | 3 - 4 | 710 | 98 | |
| XM_018040406.1 | XP_017895895.1 | 2 - 3 | 245 | 98 | ||
| GenBank mRNA | JO631860.2 | - | 2 - 3 | 265 | 98 | |
More Information
Set Membership: |
|

RAMP1
Genomic Map