Assay Details
| Gene Symbol: TMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063699.1 | XP_017919188.1 | 6 - 7 | 1348 | 55 | |
| XM_018063700.1 | XP_017919189.1 | 6 - 7 | 1376 | 55 | ||
| XM_018063701.1 | XP_017919190.1 | 6 - 7 | 1300 | 55 | ||
| XM_018063702.1 | XP_017919191.1 | 6 - 7 | 1351 | 55 | ||
| XM_018063703.1 | XP_017919192.1 | 6 - 7 | 1354 | 55 | ||
| XM_018063704.1 | XP_017919193.1 | 6 - 7 | 1357 | 55 | ||
| GenBank mRNA | JO577302.1 | - | 10 - 11 | 1841 | 55 | |
More Information
Set Membership: |
|

TMC8
Genomic Map