Assay Details
| Gene Symbol: NLRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692931.2 | XP_005692988.2 | 4 - 5 | 2175 | 62 | |
| XM_005692932.2 | XP_005692989.2 | 6 - 7 | 2175 | 62 | ||
More Information
Set Membership: |
|

NLRP9
Genomic Map