Assay Details
| Gene Symbol: KBTBD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973727.2 | XP_013829181.2 | 2 - 3 | 1763 | 67 | |
| XM_018038423.1 | XP_017893912.1 | 2 - 3 | 1347 | 67 | ||
| XM_018038424.1 | XP_017893913.1 | 2 - 3 | 1352 | 67 | ||
| XM_018038425.1 | XP_017893914.1 | 2 - 3 | 1350 | 67 | ||
| GenBank mRNA | JO380862.2 | - | 4 - 5 | 1095 | 67 | |
More Information
Set Membership: |
|

KBTBD12
Genomic Map