Assay Details
| Gene Symbol: ODAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681724.3 | XP_005681781.2 | 1 - 2 | 53 | 66 | |
| XM_013964694.2 | XP_013820148.2 | 1 - 2 | 66 | 66 | ||
More Information
Set Membership: |
|

ODAM
Genomic Map