Assay Details
Gene Symbol: HPSE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698313.2 | XP_005698370.2 | 6 - 7 | 1132 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
