Assay Details
| Gene Symbol: ZBTB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055732.1 | XP_017911221.1 | 2 | 1398 | 67 | |
| XM_018055733.1 | XP_017911222.1 | 2 | 1416 | 67 | ||
| XM_018055734.1 | XP_017911223.1 | 2 | 1378 | 67 | ||
| GenBank mRNA | JT701618.1 | - | 1 | 324 | 67 | |
More Information
Set Membership: |
|

ZBTB26
Genomic Map