Assay Details
| Gene Symbol: KIAA0753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972369.2 | XP_013827823.1 | 9 - 10 | 2592 | 57 | |
| XM_018064447.1 | XP_017919936.1 | 9 - 10 | 2084 | 57 | ||
| XM_018064448.1 | XP_017919937.1 | 9 - 10 | 2080 | 57 | ||
| XM_018064449.1 | XP_017919938.1 | 9 - 10 | 2084 | 57 | ||
| XM_018064450.1 | XP_017919939.1 | 9 - 10 | 2084 | 57 | ||
More Information
Set Membership: |
|

KIAA0753
Genomic Map