Assay Details
Gene Symbol: FAM162A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045468.1 | XP_017900957.1 | 3 - 4 | 233 | 65 | |
XM_018045498.1 | XP_017900987.1 | 2 - 3 | 265 | 65 | ||
GenBank mRNA | JO362206.2 | - | 3 - 4 | 271 | 65 | |
JO375117.2 | - | 3 - 4 | 415 | 65 | ||
JO413215.2 | - | 2 - 3 | 244 | 65 | ||
JO413216.2 | - | 3 - 4 | 433 | 65 | ||
JO449194.2 | - | 2 - 3 | 304 | 65 | ||
JO556825.2 | - | 3 - 4 | 423 | 65 | ||
JO621639.2 | - | 3 - 4 | 454 | 65 | ||
KA323098.1 | - | 2 - 3 | 270 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
