Assay Details
| Gene Symbol: STEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679305.3 | XP_005679362.1 | 4 - 5 | 1483 | 78 | |
| GenBank mRNA | JT657091.1 | - | 4 - 5 | 1675 | 78 | |
More Information
Set Membership: |
|

STEAP2
Genomic Map