Assay Details
| Gene Symbol: ZNF143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689704.3 | XP_005689761.1 | 15 - 16 | 1930 | 57 | |
| XM_005689705.3 | XP_005689762.1 | 15 - 16 | 1927 | 57 | ||
| XM_005689706.3 | XP_005689763.1 | 15 - 16 | 1954 | 57 | ||
| XM_005689707.3 | XP_005689764.1 | 15 - 16 | 1951 | 57 | ||
More Information
Set Membership: |
|

ZNF143
Genomic Map