Assay Details
| Gene Symbol: LIMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065343.1 | XP_017920832.1 | 3 - 4 | 281 | 68 | |
| GenBank mRNA | JO341414.2 | - | 2 - 3 | 1051 | 68 | |
More Information
Set Membership: |
|

LIMD2
Genomic Map