Assay Details
| Gene Symbol: FAM222B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693267.3 | XP_005693324.2 | 2 - 3 | 406 | 96 | |
| XM_018064321.1 | XP_017919810.1 | 2 - 3 | 263 | 96 | ||
More Information
Set Membership: |
|

FAM222B
Genomic Map