Assay Details
| Gene Symbol: MIEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964357.2 | XP_013819811.2 | 4 - 5 | 862 | 73 | |
| XM_018048792.1 | XP_017904281.1 | 4 - 5 | 861 | 73 | ||
| GenBank mRNA | JO426092.1 | - | 4 - 5 | 912 | 73 | |
| JO431891.2 | - | 4 - 5 | 852 | 73 | ||
More Information
Set Membership: |
|

MIEF1
Genomic Map