Assay Details
| Gene Symbol: MB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680603.3 | XP_005680660.1 | 3 - 4 | 369 | 74 | |
| XM_013964067.2 | XP_013819521.1 | 4 - 5 | 491 | 74 | ||
| GenBank mRNA | FJ457764.1 | - | 2 - 3 | 273 | 74 | |
| JO537826.2 | - | 1 - 2 | 851 | 74 | ||
More Information
Set Membership: |
|

MB
Genomic Map