Assay Details
| Gene Symbol: RRP7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048859.1 | XP_017904348.1 | 1 - 2 | 97 | 86 | |
| GenBank mRNA | JO584944.2 | - | 1 - 2 | 126 | 86 | |
| JO584945.2 | - | 1 - 2 | 126 | 86 | ||
More Information
Set Membership: |
|

RRP7A
Genomic Map