Assay Details
Gene Symbol: ST6GALNAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055833.1 | XP_017911322.1 | 3 - 4 | 435 | 107 | |
XM_018055835.1 | XP_017911324.1 | 3 - 4 | 606 | 107 | ||
XM_018055836.1 | XP_017911325.1 | 3 - 4 | 435 | 107 | ||
XM_018055837.1 | XP_017911326.1 | 3 - 4 | 435 | 107 | ||
XM_018055838.1 | XP_017911327.1 | 3 - 4 | 435 | 107 | ||
GenBank mRNA | JO343908.2 | - | 2 - 3 | 261 | 107 | |
JO433997.2 | - | 2 - 3 | 316 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
