Assay Details
| Gene Symbol: BMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057630.1 | XP_017913119.1 | 6 - 7 | 1668 | 58 | |
| GenBank mRNA | JO393146.2 | - | 6 - 7 | 1678 | 58 | |
More Information
Set Membership: |
|

BMP7
Genomic Map