Assay Details
| Gene Symbol: RRP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690476.3 | XP_005690533.3 | 4 - 5 | 764 | 70 | |
| GenBank mRNA | JO585333.2 | - | 1 - 2 | 243 | 70 | |
| KA309982.1 | - | 1 - 2 | 312 | 70 | ||
More Information
Set Membership: |
|

RRP15
Genomic Map